siRNA Details
| Gene Symbol: Cyp27a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024264.4 | NP_077226.2 | 2 | 353 | ||
| GenBank mRNA | AK004977.1 | - | 2 | 354 | ||
| BC002183.1 | - | 1 | 27 | 3487429 | ||
| BC055028.1 | - | 2 | 331 | 5098366 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp27a1| Gene Symbol: Cyp27a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024264.4 | NP_077226.2 | 2 | 353 | ||
| GenBank mRNA | AK004977.1 | - | 2 | 354 | ||
| BC002183.1 | - | 1 | 27 | 3487429 | ||
| BC055028.1 | - | 2 | 331 | 5098366 | ||