siRNA Details
| Gene Symbol: Gldc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138595.2 | NP_613061.1 | 25 | 3217 | ||
| GenBank mRNA | AK080789.1 | - | 25 | 3189 | ||
| AK080937.1 | - | 25 | 3207 | |||
| BC017135.1 | - | 25 | 3194 | 4511573 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gldc| Gene Symbol: Gldc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138595.2 | NP_613061.1 | 25 | 3217 | ||
| GenBank mRNA | AK080789.1 | - | 25 | 3189 | ||
| AK080937.1 | - | 25 | 3207 | |||
| BC017135.1 | - | 25 | 3194 | 4511573 | ||