siRNA Details
| Gene Symbol: Gas8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018855.2 | NP_061343.2 | 9, 10 | 1344 | ||
| GenBank mRNA | AK140135.1 | - | 9, 10 | 1322 | ||
| AY692443.1 | - | 9, 10 | 1343 | |||
| BC003779.1 | - | 9, 10 | 1259 | 3592695 | ||
| U19859.1 | - | 9, 10 | 1365 | |||

Gas8
Genomic Map