siRNA Details
| Gene Symbol: Pcnxl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144868.3 | NP_659117.2 | 2 | 1034 | ||
| GenBank mRNA | AF237953.1 | - | 2 | 275 | ||
| AK171516.1 | - | 2 | 821 | |||
| BC054751.1 | - | 2 | 313 | |||
| BC055289.1 | - | 3 | 837 | |||
| BC096538.1 | - | 2 | 1034 | |||

Pcnxl3
Genomic Map