siRNA Details
| Gene Symbol: Bap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027088.2 | NP_081364.1 | 13 | 1406 | ||
| GenBank mRNA | AB047820.1 | - | 13 | 1307 | ||
| AK079374.1 | - | 13 | 1424 | |||
| AK129108.1 | - | 13 | 1314 | |||
| AK170576.1 | - | 13 | 1407 | |||
| AK171561.1 | - | 13 | 1409 | |||
| BC042489.1 | - | 12 | 1854 | |||
| BC050901.1 | - | 13 | 1369 | 6402443 | ||

Bap1
Genomic Map