siRNA Details
| Gene Symbol: Bap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027088.2 | NP_081364.1 | 14, 15 | 2022 | ||
| GenBank mRNA | AB047820.1 | - | 14, 15 | 1923 | ||
| AK009033.1 | - | 1, 2 | 36 | |||
| AK079374.1 | - | 14, 15 | 2040 | |||
| AK129108.1 | - | 14, 15 | 1930 | |||
| AK170576.1 | - | 14, 15 | 2023 | |||
| AK171561.1 | - | 14, 15 | 2025 | |||
| BC042489.1 | - | 13, 14 | 2470 | |||
| BC050901.1 | - | 14, 15 | 1985 | 6402443 | ||

Bap1
Genomic Map