siRNA Details
| Gene Symbol: Mycbpap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170671.2 | NP_733771.2 | 8 | 990 | ||
| GenBank mRNA | AK029929.1 | - | 7 | 1166 | ||
| AK166780.1 | - | 1 | 138 | |||
| BC027327.1 | - | 4 | 351 | 3991131 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mycbpap| Gene Symbol: Mycbpap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170671.2 | NP_733771.2 | 8 | 990 | ||
| GenBank mRNA | AK029929.1 | - | 7 | 1166 | ||
| AK166780.1 | - | 1 | 138 | |||
| BC027327.1 | - | 4 | 351 | 3991131 | ||