siRNA Details
| Gene Symbol: Tsr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177325.3 | NP_796299.2 | 5 | 959 | ||
| GenBank mRNA | AK030545.1 | - | 4 | 865 | ||
| AK129350.1 | - | 5 | 913 | |||
| AK134365.1 | - | 5 | 943 | |||
| AK154063.1 | - | 5 | 946 | |||
| BC031531.1 | - | 5 | 932 | 4014819 | ||

Tsr1
Genomic Map