siRNA Details
| Gene Symbol: Slc16a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029842.1 | NP_001025013.1 | 5 | 1301 | ||
| NM_134038.2 | NP_598799.1 | 5 | 1244 | |||
| GenBank mRNA | AK032026.1 | - | 5 | 1304 | ||
| AK141927.1 | - | 1 | 1184 | |||
| AK170472.1 | - | 5 | 1167 | |||
| BC017129.1 | - | 5 | 1235 | 4506255 | ||

Slc16a6
Genomic Map