siRNA Details
| Gene Symbol: Pld4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178911.4 | NP_849242.1 | 7 | 941 | ||
| GenBank mRNA | AK045459.1 | - | 7 | 900 | ||
| AK079549.1 | - | 7 | 900 | |||
| AK079878.1 | - | 7 | 943 | |||
| AK151551.1 | - | 7 | 1175 | |||
| AK152969.1 | - | 6 | 788 | |||
| AK170362.1 | - | 7 | 898 | |||
| BC058565.1 | - | 7 | 891 | 6442038 | ||

Pld4
Genomic Map