siRNA Details
| Gene Symbol: Rab15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134050.4 | NP_598811.3 | 7 | 737 | ||
| GenBank mRNA | AB232610.1 | - | 7 | 530 | ||
| AK158637.1 | - | 7 | 659 | |||
| AK158768.1 | - | 7 | 691 | |||
| BC027769.1 | - | 7 | 737 | 5345297 | ||

Rab15
Genomic Map