siRNA Details
| Gene Symbol: Arrdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042591.1 | NP_001036056.1 | 6 | 1128 | ||
| GenBank mRNA | AK084951.1 | - | 6 | 1139 | ||
| AK088288.1 | - | 6 | 1143 | |||
| AK122499.1 | - | 6 | 1115 | |||
| AK163364.1 | - | 6 | 673 | |||
| BC023838.1 | - | 3 | 346 | |||
| BC031125.1 | - | 4 | 506 | |||
| BC044762.1 | - | 6 | 1126 | |||
| BC054844.1 | - | 6 | 1128 | 6493441 | ||
| BC098474.1 | - | 6 | 1122 | 30094818 | ||
| DQ861994.1 | - | 6 | 873 | |||

Arrdc3
Genomic Map