siRNA Details
| Gene Symbol: Golm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035122.2 | NP_001030294.1 | 9 | 1131 | ||
| NM_027307.4 | NP_081583.3 | 10 | 1267 | |||
| GenBank mRNA | AK009072.1 | - | 8 | 938 | ||
| AK133865.1 | - | 9 | 1133 | |||
| AK167646.1 | - | 9 | 1108 | |||
| BC011152.1 | - | 9 | 1092 | 4188434 | ||
| BC098486.1 | - | 10 | 1263 | 30024953 | ||
| CN529028.1 | - | 1 | 69 | |||

Golm1
Genomic Map