siRNA Details
| Gene Symbol: Golm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035122.2 | NP_001030294.1 | 7 | 789 | ||
| NM_027307.4 | NP_081583.3 | 8 | 925 | |||
| GenBank mRNA | AK009072.1 | - | 6 | 596 | ||
| AK133865.1 | - | 7 | 791 | |||
| AK167646.1 | - | 7 | 766 | |||
| BC011152.1 | - | 7 | 750 | 4188434 | ||

Golm1
Genomic Map