siRNA Details
| Gene Symbol: Dusp22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037955.4 | NP_001033044.1 | 6 | 358 | ||
| NM_134068.3 | NP_598829.1 | 6 | 358 | |||
| GenBank mRNA | AF237619.1 | - | 6 | 318 | ||
| AK148708.1 | - | 6 | 326 | |||
| AK149363.1 | - | 6 | 317 | |||
| BC080718.1 | - | 6 | 308 | |||
| BC108362.1 | - | 6 | 308 | 4917645 | ||
| CX236611.1 | - | 6 | 358 | |||

Dusp22
Genomic Map