siRNA Details
| Gene Symbol: Akr1c14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134072.1 | NP_598833.1 | 1, 2 | 119 | ||
| GenBank mRNA | AK033537.1 | - | 1, 2 | 137 | ||
| AK136528.1 | - | 1, 2 | 136 | |||
| AY730283.1 | - | 1, 2 | 81 | |||
| BC013482.1 | - | 1, 2 | 119 | 4240772 | ||
| BC026628.1 | - | 1, 2 | 118 | 4983237 | ||

Akr1c14
Genomic Map