siRNA Details
Gene Symbol: E330034G19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033214.2 | NP_001028386.1 | 5 | 682 | ||
GenBank mRNA | AK136036.1 | - | 5 | 683 | ||
AK163316.1 | - | 5 | 474 | |||
BC139082.1 | - | 5 | 653 | 8862104 | ||
BC139083.1 | - | 5 | 653 | 8862105 |
