siRNA Details
| Gene Symbol: Dock9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081039.1 | NP_001074508.1 | 31 | 3512 | ||
| NM_001128307.1 | NP_001121779.1 | 31 | 3917 | |||
| NM_001128308.1 | NP_001121780.1 | 31 | 3917 | |||
| NM_134074.2 | NP_598835.2 | 31 | 3533 | |||
| GenBank mRNA | AK122431.1 | - | 31 | 3916 | ||
| BC046250.1 | - | 25 | 2819 | |||
| BC052947.1 | - | 25 | 2831 | |||
| BC157964.1 | - | 31 | 3457 | 9007289 | ||
| BC158050.1 | - | 31 | 3848 | 9007354 | ||
| BC158127.1 | - | 31 | 3416 | |||

Dock9
Genomic Map