siRNA Details
| Gene Symbol: Mmrn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153127.3 | NP_694767.3 | 2 | 316 | ||
| GenBank mRNA | AK077884.1 | - | 2 | 318 | ||
| AK078470.1 | - | 2 | 292 | |||
| AK134285.1 | - | 2 | 356 | |||
| BC034871.1 | - | 2 | 258 | |||
| BC145845.1 | - | 2 | 236 | 40131148 | ||

Mmrn2
Genomic Map