siRNA Details
| Gene Symbol: Mtbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168250.1 | NP_001161722.1 | 11, 12 | 1355 | ||
| NM_134092.3 | NP_598853.3 | 11, 12 | 1355 | |||
| GenBank mRNA | AJ278508.1 | - | 11, 12 | 1276 | ||
| AK080036.1 | - | 11, 12 | 1169 | |||
| AK088549.1 | - | 11, 12 | 1169 | |||
| AK088652.1 | - | 1, 2 | 783 | |||
| AK133855.1 | - | 11, 12 | 1203 | |||
| BC005500.1 | - | 4, 5 | 352 | |||
| BC052197.1 | - | 11, 12 | 1355 | 6511752 | ||

Mtbp
Genomic Map