siRNA Details
| Gene Symbol: Dennd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081066.1 | NP_001074535.1 | 20 | 3486 | ||
| GenBank mRNA | AK053114.1 | - | 10 | 1820 | ||
| AK173046.1 | - | 20 | 3386 | |||
| BC132389.1 | - | 20 | 3374 | 40130666 | ||
| BC137791.1 | - | 20 | 3374 | 8860810 | ||
| BC144912.1 | - | 20 | 3374 | |||

Dennd3
Genomic Map