siRNA Details
| Gene Symbol: Topors | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134097.3 | NP_598858.2 | 3 | 1077 | ||
| GenBank mRNA | AB072395.1 | - | 3 | 1034 | ||
| AB104865.1 | - | 3 | 1062 | |||
| AK044564.1 | - | 3 | 1055 | |||
| AK134075.1 | - | 3 | 1073 | |||
| AK140250.1 | - | 3 | 1078 | |||
| AK143025.1 | - | 3 | 1058 | |||
| AK153743.1 | - | 3 | 1064 | |||
| BC019421.1 | - | 3 | 1035 | |||
| BC037141.1 | - | 3 | 1055 | |||
| BC040797.1 | - | 3 | 1065 | 3492178 | ||

Topors
Genomic Map