siRNA Details
| Gene Symbol: Gga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145929.2 | NP_666041.1 | 15 | 1944 | ||
| GenBank mRNA | AK080881.1 | - | 15 | 1567 | ||
| AK155195.1 | - | 15 | 1584 | |||
| AK159771.1 | - | 15 | 1567 | |||
| AK170437.1 | - | 15 | 1583 | |||
| AK189555.1 | - | 2 | 136 | |||
| AK194733.1 | - | 2 | 136 | |||
| BC026802.1 | - | 15 | 1944 | 4223300 | ||
| BC054422.1 | - | 15 | 1421 | |||

Gga1
Genomic Map