siRNA Details
| Gene Symbol: Gga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145929.2 | NP_666041.1 | 11 | 1455 | ||
| GenBank mRNA | AK080881.1 | - | 11 | 1078 | ||
| AK155195.1 | - | 11 | 1095 | |||
| AK159771.1 | - | 11 | 1078 | |||
| AK170437.1 | - | 11 | 1092 | |||
| AK171534.1 | - | 11 | 1112 | |||
| BC026802.1 | - | 11 | 1455 | 4223300 | ||
| BC054422.1 | - | 11 | 1099 | |||

Gga1
Genomic Map