siRNA Details
| Gene Symbol: Gga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145929.2 | NP_666041.1 | 10 | 1239 | ||
| GenBank mRNA | AK080881.1 | - | 10 | 862 | ||
| AK155195.1 | - | 10 | 879 | |||
| AK159771.1 | - | 10 | 862 | |||
| AK170437.1 | - | 10 | 880 | |||
| AK171534.1 | - | 10 | 896 | |||
| AK185927.1 | - | 3 | 215 | |||
| AK188934.1 | - | 3 | 216 | |||
| AK210879.1 | - | 3 | 215 | |||
| AK218460.1 | - | 3 | 215 | |||
| BC026802.1 | - | 10 | 1239 | 4223300 | ||
| BC054422.1 | - | 10 | 883 | |||

Gga1
Genomic Map