siRNA Details
| Gene Symbol: Qtrtd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029128.2 | NP_083404.2 | 5 | 627 | ||
| GenBank mRNA | AK014045.1 | - | 6 | 985 | ||
| AK015536.1 | - | 878 | ||||
| BC017628.1 | - | 6 | 812 | |||
| BC083089.1 | - | 5 | 607 | 30105859 | ||
| FM985972.1 | - | 5 | 720 | |||

Qtrtd1
Genomic Map