siRNA Details
| Gene Symbol: Ldhal6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175349.2 | NP_780558.1 | 1 | 900 | ||
| GenBank mRNA | AK077089.1 | - | 1 | 846 | ||
| AY280363.1 | - | 1 | 900 | |||
| BC061193.1 | - | 1 | 845 | 6704800 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ldhal6b| Gene Symbol: Ldhal6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175349.2 | NP_780558.1 | 1 | 900 | ||
| GenBank mRNA | AK077089.1 | - | 1 | 846 | ||
| AY280363.1 | - | 1 | 900 | |||
| BC061193.1 | - | 1 | 845 | 6704800 | ||