siRNA Details
| Gene Symbol: Rpusd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028009.3 | NP_082285.1 | 6 | 670 | ||
| GenBank mRNA | AK009926.1 | - | 5 | 502 | ||
| AK049334.1 | - | 6 | 671 | |||
| BC018186.1 | - | 6 | 631 | 4160116 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rpusd1| Gene Symbol: Rpusd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028009.3 | NP_082285.1 | 6 | 670 | ||
| GenBank mRNA | AK009926.1 | - | 5 | 502 | ||
| AK049334.1 | - | 6 | 671 | |||
| BC018186.1 | - | 6 | 631 | 4160116 | ||