siRNA Details
| Gene Symbol: Ticam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174989.4 | NP_778154.1 | 2 | 1369 | ||
| GenBank mRNA | AB091053.1 | - | 1 | 1236 | ||
| AK054219.1 | - | 2 | 1329 | |||
| AK083836.1 | - | 2 | 1347 | |||
| AK086237.1 | - | 2 | 1347 | |||
| AK089679.1 | - | 2 | 1347 | |||
| AK143766.1 | - | 2 | 1361 | |||
| AK150150.1 | - | 2 | 1363 | |||
| AK155245.1 | - | 2 | 1369 | |||
| AK155906.1 | - | 2 | 1361 | |||
| AK171578.1 | - | 2 | 1361 | |||
| BC033406.1 | - | 1 | 124 | |||
| BC037048.1 | - | 2 | 1369 | |||
| BC094338.1 | - | 1 | 1305 | 30606722 | ||

Ticam1
Genomic Map