siRNA Details
| Gene Symbol: Dhx57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163759.1 | NP_001157231.1 | 17 | 3509 | ||
| NM_198942.2 | NP_945180.2 | 16 | 3350 | |||
| GenBank mRNA | AK035344.1 | - | 1 | 191 | ||
| BC026474.1 | - | 4 | 491 | |||
| BC037692.1 | - | 5 | 586 | |||
| BC062952.1 | - | 16 | 3350 | 6853031 | ||
| BC065169.1 | - | 17 | 3176 | 6831314 | ||
| BC066091.1 | - | 16 | 3012 | 6841207 | ||
| BC079618.1 | - | 16 | 3630 | |||

Dhx57
Genomic Map