siRNA Details
| Gene Symbol: Oard1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289490.1 | NP_001276419.1 | 2, 3 | 443 | ||
| NM_001289491.1 | NP_001276420.1 | 2, 3 | 168 | |||
| NM_207219.4 | NP_997102.2 | 2, 3 | 308 | |||
| GenBank mRNA | AK148136.1 | - | 2, 3 | 430 | ||
| AK153512.1 | - | 2, 3 | 231 | |||
| BC022574.1 | - | 1, 2 | 87 | 3989373 | ||
| BC157987.1 | - | 2, 3 | 193 | 9007303 | ||
| BY741306.1 | - | 2, 3 | 430 | |||

Oard1
Genomic Map