siRNA Details
| Gene Symbol: Arap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205336.1 | NP_001192265.1 | 21, 22 | 3376 | ||
| NM_139206.2 | NP_631945.2 | 21, 22 | 3176 | |||
| GenBank mRNA | AF469621.1 | - | 20, 21 | 3126 | ||
| AF469622.1 | - | 21, 22 | 3317 | |||
| AK220486.1 | - | 20, 21 | 3605 | |||
| BC068145.1 | - | 21, 22 | 3164 | 6856630 | ||

Arap3
Genomic Map