siRNA Details
| Gene Symbol: Fbxo38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134136.3 | NP_598897.2 | 6, 7 | 907 | ||
| GenBank mRNA | AK031096.1 | - | 6, 7 | 857 | ||
| AK031347.1 | - | 6, 7 | 838 | |||
| AK034761.1 | - | 1, 2 | 91 | |||
| AK089046.1 | - | 6, 7 | 879 | |||
| AY267463.1 | - | 6, 7 | 840 | |||
| BC056348.1 | - | 6, 7 | 859 | 5721407 | ||

Fbxo38
Genomic Map