siRNA Details
Gene Symbol: Btaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080706.1 | NP_001074175.1 | 18 | 2509 | ||
GenBank mRNA | AB287295.1 | - | 18 | 2234 | ||
AK141888.1 | - | 12 | 2956 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Btaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080706.1 | NP_001074175.1 | 18 | 2509 | ||
GenBank mRNA | AB287295.1 | - | 18 | 2234 | ||
AK141888.1 | - | 12 | 2956 |