siRNA Details
| Gene Symbol: Otub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134150.2 | NP_598911.1 | 2 | 121 | ||
| GenBank mRNA | AK027996.1 | - | 2 | 108 | ||
| AK087195.1 | - | 2 | 108 | |||
| AK140070.1 | - | 2 | 108 | |||
| AK145273.1 | - | 2 | 111 | |||
| AK145970.1 | - | 2 | 113 | |||
| BC022575.1 | - | 2 | 121 | 5005025 | ||
| BC054410.1 | - | 2 | 89 | |||

Otub1
Genomic Map