siRNA Details
| Gene Symbol: Yars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134151.4 | NP_598912.4 | 6 | 861 | ||
| GenBank mRNA | AK028785.1 | - | 6 | 723 | ||
| AK043837.1 | - | 6 | 785 | |||
| AK076634.1 | - | 6 | 706 | |||
| AK139579.1 | - | 6 | 706 | |||
| AK145356.1 | - | 6 | 754 | |||
| AK151880.1 | - | 6 | 713 | |||
| AK169708.1 | - | 6 | 637 | |||
| AK177997.1 | - | 2 | 140 | |||
| BC013552.1 | - | 6 | 792 | 4236514 | ||
| BC022143.1 | - | 6 | 756 | 5101324 | ||
| BC026615.1 | - | 6 | 768 | 4976999 | ||

Yars
Genomic Map