siRNA Details
| Gene Symbol: Yars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134151.4 | NP_598912.4 | 4 | 698 | ||
| GenBank mRNA | AK028785.1 | - | 4 | 560 | ||
| AK043837.1 | - | 4 | 622 | |||
| AK076634.1 | - | 4 | 543 | |||
| AK139579.1 | - | 4 | 543 | |||
| AK145356.1 | - | 4 | 591 | |||
| AK151880.1 | - | 4 | 550 | |||
| AK169708.1 | - | 4 | 474 | |||
| BC013552.1 | - | 4 | 629 | 4236514 | ||
| BC022143.1 | - | 4 | 593 | 5101324 | ||
| BC026615.1 | - | 4 | 605 | 4976999 | ||

Yars
Genomic Map