siRNA Details
| Gene Symbol: Lpxn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134152.3 | NP_598913.1 | 2, 3 | 239 | ||
| GenBank mRNA | AB053936.2 | - | 2, 3 | 291 | ||
| AB071194.1 | - | 2, 3 | 264 | |||
| AK077085.1 | - | 2, 3 | 275 | |||
| BC026563.1 | - | 2, 3 | 205 | 5065405 | ||

Lpxn
Genomic Map