siRNA Details
| Gene Symbol: Gbf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178930.3 | NP_849261.2 | 31 | 4245 | ||
| GenBank mRNA | AK172919.1 | - | 28 | 3838 | ||
| BC023791.1 | - | 5 | 663 | |||
| BC024129.1 | - | 14 | 1903 | |||
| BC026621.1 | - | 2 | 277 | |||
| BC034745.1 | - | 16 | 2010 | |||
| BC059883.1 | - | 8 | 910 | |||
| BC060728.1 | - | 23 | 3665 | |||
| BC076569.1 | - | 31 | 4245 | 6409170 | ||
| BC082336.1 | - | 32 | 4240 | |||

Gbf1
Genomic Map