siRNA Details
| Gene Symbol: Eprs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029735.1 | NP_084011.1 | 15 | 1924 | ||
| GenBank mRNA | AK028687.1 | - | 8 | 945 | ||
| AK141072.1 | - | 15 | 1979 | |||
| AK148463.1 | - | 15 | 1885 | |||
| AK160986.1 | - | 15 | 1945 | |||
| BC003226.1 | - | 3 | 241 | |||
| BC040802.1 | - | 15 | 1922 | |||
| BC058337.1 | - | 16 | 1898 | |||
| BC094679.1 | - | 14 | 1718 | |||
| BC141049.1 | - | 15 | 1843 | 9055661 | ||
| X54327.1 | - | 6 | 608 | |||

Eprs
Genomic Map