siRNA Details
| Gene Symbol: Osr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054049.2 | NP_473390.1 | 2, 3 | 1124 | ||
| GenBank mRNA | AF370121.1 | - | 2, 3 | 1124 | ||
| AK052147.1 | - | 2, 3 | 1139 | |||
| AY038074.1 | - | 1, 2 | 657 | |||
| BC013504.1 | - | 2, 3 | 931 | 4224339 | ||

Osr2
Genomic Map