siRNA Details
Gene Symbol: Mylk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139300.3 | NP_647461.3 | 20 | 3912 | ||
GenBank mRNA | AK078556.1 | - | 5 | 1351 | ||
AK162008.1 | - | 2 | 278 | |||
AY237726.1 | - | 20 | 3941 | |||
AY237727.2 | - | 5 | 1465 | |||
BC030478.1 | - | 10 | 2144 | |||
BC037721.1 | - | 5 | 475 | |||
BC045197.1 | - | 5 | 1395 | |||
BC058610.1 | - | 20 | 3924 | 30056142 | ||
BC145279.1 | - | 20 | 3899 | 9053833 | ||
BC150749.1 | - | 20 | 3899 | 9087660 | ||
BC171954.1 | - | 20 | 3900 |
Gene Symbol: E130310I04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK044449.1 | - | 3 | 1876 | ||
AK053822.1 | - | 3 | 1863 |
