siRNA Details
| Gene Symbol: Nod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171007.1 | NP_001164478.1 | 4 | 1881 | ||
| NM_172729.3 | NP_766317.1 | 4 | 2132 | |||
| GenBank mRNA | AK082663.1 | - | 1 | 927 | ||
| AK089662.1 | - | 4 | 1882 | |||
| AK137585.1 | - | 4 | 2137 | |||
| AY160222.1 | - | 3 | 1667 | |||
| BC042670.1 | - | 4 | 1959 | 3597911 | ||
| BC043670.1 | - | 1 | 849 | |||

Nod1
Genomic Map