siRNA Details
| Gene Symbol: Nod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171007.1 | NP_001164478.1 | 2 | 328 | ||
| NM_172729.3 | NP_766317.1 | 2 | 579 | |||
| GenBank mRNA | AK089662.1 | - | 2 | 329 | ||
| AK137585.1 | - | 2 | 584 | |||
| AY160222.1 | - | 1 | 114 | |||
| BC042670.1 | - | 2 | 406 | 3597911 | ||

Nod1
Genomic Map