siRNA Details
| Gene Symbol: Uap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036243.1 | - | 4 | 783 | ||
| AK047566.1 | - | 4 | 848 | |||
| AK049519.1 | - | 4 | 837 | |||
| AK081690.1 | - | 4 | 808 | |||
| AK147139.1 | - | 4 | 783 | |||
| AK163314.1 | - | 4 | 682 | |||
| AK171925.1 | - | 4 | 680 | |||
| BC016406.1 | - | 4 | 770 | 4489112 | ||
| BC017547.1 | - | 4 | 659 | 4920702 | ||

Uap1
Genomic Map