siRNA Details
| Gene Symbol: Slc12a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133648.2 | NP_598409.2 | 9 | 1083 | ||
| NM_133649.2 | NP_598410.2 | 9 | 1744 | |||
| GenBank mRNA | AF211854.1 | - | 9 | 1239 | ||
| AF211855.1 | - | 9 | 1083 | |||
| AK133006.1 | - | 9 | 2234 | |||
| AK137387.1 | - | 8 | 1063 | |||
| BC062099.1 | - | 1699 | 6844732 | |||

Slc12a6
Genomic Map