siRNA Details
| Gene Symbol: Tm6sf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291282.1 | NP_001278211.1 | 7 | 579 | ||
| NM_145375.4 | NP_663350.2 | 7 | 1087 | |||
| GenBank mRNA | AK083132.1 | - | 7 | 1090 | ||
| AK135100.1 | - | 6 | 3309 | |||
| AK136581.1 | - | 7 | 580 | |||
| AK163621.1 | - | 7 | 1012 | |||
| AK172109.1 | - | 7 | 1003 | |||
| AK172145.1 | - | 7 | 988 | |||
| BC023123.1 | - | 7 | 969 | 3995280 | ||

Tm6sf1
Genomic Map