siRNA Details
| Gene Symbol: Tm6sf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293795.1 | NP_001280724.1 | 8, 9 | 873 | ||
| NM_181540.4 | NP_853518.1 | 8, 9 | 867 | |||
| GenBank mRNA | AK077126.1 | - | 8, 9 | 860 | ||
| BC024498.1 | - | 8, 9 | 861 | 4982620 | ||

Tm6sf2
Genomic Map