siRNA Details
| Gene Symbol: Mgat5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145128.3 | NP_660110.2 | 15 | 2362 | ||
| GenBank mRNA | AF474154.1 | - | 13 | 1742 | ||
| AF474155.1 | - | 13 | 1805 | |||
| AK030402.1 | - | 15 | 2330 | |||
| BC125533.1 | - | 13 | 1770 | 40130048 | ||
| BC125535.1 | - | 13 | 1770 | 40130050 | ||

Mgat5
Genomic Map