siRNA Details
| Gene Symbol: Pom121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148932.2 | NP_683734.2 | 2 | 629 | ||
| GenBank mRNA | AF516680.1 | - | 2 | 593 | ||
| AK154473.1 | - | 2 | 630 | |||
| BC053101.1 | - | 2 | 605 | 6405486 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pom121| Gene Symbol: Pom121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148932.2 | NP_683734.2 | 2 | 629 | ||
| GenBank mRNA | AF516680.1 | - | 2 | 593 | ||
| AK154473.1 | - | 2 | 630 | |||
| BC053101.1 | - | 2 | 605 | 6405486 | ||